Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1beta-hydroxyalantolactone | 68776-47-6 | 99.9% | 248.32 | C15H20O3 | 5MG
SDP

Catalog No. 5000236452
Encompass_Preferred

1beta-Hydroxyalantolactone is a sesquiterpene lactone research compound reported to modulate inflammatory signaling, including interleukin-related pathways and NF-κB, and is used in vitro and in vivo investigations of immunology and inflammation mechanisms.

  • Sesquiterpene lactone reported to modulate interleukin-related signaling and NF-κB.
  • CAS No. 68776-47-6.
  • Formula C15H20O3; molecular weight 248.32.
  • Purity 99.94%.
  • Appearance white to off-white solid.
  • Solubility: DMSO 50 mg/mL (201.35 mM; ultrasonic recommended); in vivo ≥ 1.25 mg/mL (5.03 mM), clear solution.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Pack size 5 mg, suitable for small-scale biological research applications.

Catalog No. 50-002-36452 Supplier Medchemexpress LLC Supplier No. HY1246705MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1beta-Hydroxyalantolactone is a sesquiterpene lactone research compound reported to modulate inflammatory signaling, including interleukin-related pathways and NF-κB, and is used in vitro and in vivo investigations of immunology and inflammation mechanisms.

  • Sesquiterpene lactone reported to modulate interleukin-related signaling and NF-κB.
  • CAS No. 68776-47-6.
  • Formula C15H20O3; molecular weight 248.32.
  • Purity 99.94%.
  • Appearance white to off-white solid.
  • Solubility: DMSO 50 mg/mL (201.35 mM; ultrasonic recommended); in vivo ≥ 1.25 mg/mL (5.03 mM), clear solution.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Pack size 5 mg, suitable for small-scale biological research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.