Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Butyl-3-Methylimidazolium Acetate

Catalog No. 399521KGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-952-1KGF 1 kg
39-952-100GF 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-952-1KGF Supplier Sigma Aldrich Supplier No. 399521KGF
Only null left
Add to cart
Add to cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Butyl-3-methylimidazolium acetate is a 1-alkyl-3-methyl imidazolium based ionic liquid.1-Butyl-3-methylimidazolium acetate can be used as an additive to electrolytes for non-aqueous capillary electrophoresis (NACE) due to its air- and water-stability and miscibility with most of CE solvents.

Specifications

CAS 284049-75-8
Molecular Formula C10H18N2O2
Linear Formula C10H18N2O2
MDL Number MFCD06798175
Quantity 1 kg
Molecular Weight (g/mol) 198.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.