Learn More
Cayman Chemical 1-Carboxycyclohexaneacetic Acid, Purity: ≥95%, CAS: 67950-95-2, 50 mg
SDP
Catalog No. 502911936
Encompass_Preferred
A potential impurity in commercial preparations of gabapentin; has been used as a precursor in the synthesis of 5-HT2A receptor antagonists
Catalog No. 50-291-1936 Supplier Cayman Chemical Supplier No. 3604650 MG
A potential impurity in commercial preparations of gabapentin; has been used as a precursor in the synthesis of 5-HT2A receptor antagonists