Learn More
DUTCH OPHTHALMIC USA 1cc syringe holder for micro-injection, Box/6
Catalog No. NC3706126
Encompass
EVA INICIO™: developed for EVA NEXUS™, the first approved ophthalmic microinjection system. Precise delivery and control of microinjections. • Suitable for sub 1 ml microinjections • Pressure delivery of ±1 PSI • Full surgeon control by footswitch Compatible with DORC subretinal cannula 1270.06, 1270.04 and 1270.EXT.
This item is not returnable. View return policy
Catalog No. NC3706126 Supplier DUTCH OPHTHALMIC USA Supplier No. 1364.DD
EVA INICIO™: developed for EVA NEXUS™, the first approved ophthalmic microinjection system. Precise delivery and control of microinjections. • Suitable for sub 1 ml microinjections • Pressure delivery of ±1 PSI • Full surgeon control by footswitch Compatible with DORC subretinal cannula 1270.06, 1270.04 and 1270.EXT.