Learn More
Andwin Scientific 1CC SYRINGE RUB/FREE 100PK
SDP
Catalog No. NC9517067
Encompass_Preferred
The Norm-Ject� syringes are free of contaminants associated with disposables, latex, rubber, silicone oil, or styrene and are sterile. This product is ideal for sampling, mixing, and filtering. The syringes feature a positive safety stop to avoid accidental spills. This product has accurate and easily readable 0.01 mL graduation increments and a volume capacity of 1 mL. It is sold in a pack of 100.
This item is not returnable. View return policy
Catalog No. NC9517067 Supplier Andwin Scientific Supplier No. 53548000
 Restricted Item
Add to Cart
Add to Cart
The Norm-Ject� syringes are free of contaminants associated with disposables, latex, rubber, silicone oil, or styrene and are sterile. This product is ideal for sampling, mixing, and filtering. The syringes feature a positive safety stop to avoid accidental spills. This product has accurate and easily readable 0.01 mL graduation increments and a volume capacity of 1 mL. It is sold in a pack of 100.