Learn More
Cayman Chemical 1-β-D-2'-Deoxyribofuranosyl-1,2,4-triazole-3-carboxamide-5'-O-triphosphate (sodium salt), Purity: ≥95%, 2.3 mg
SDP
Catalog No. 502919132
Encompass_Preferred
A nucleotide analog; has been used in SeSaM for the generation of mutant libraries
Catalog No. 50-291-9132 Supplier Cayman Chemical Supplier No. 385622.3 MG
A nucleotide analog; has been used in SeSaM for the generation of mutant libraries