Learn More
Cayman Chemical 1-β-D-Arabinofuranosyluracil, Purity: ≥98%, CAS: 3083-77-0, 10 g
SDP
Catalog No. 502990142
Encompass_Preferred
An inactive metabolite of cytarabine; formed when cytarabine undergoes deamination by cytidine deaminase
Catalog No. 50-299-0142 Supplier Cayman Chemical Supplier No. 2987510 G
An inactive metabolite of cytarabine; formed when cytarabine undergoes deamination by cytidine deaminase