Learn More
Cayman Chemical 1DeoxygalactonjIrImycInhy 10mg
SDP
Catalog No. 502968457
Encompass_Preferred
A competitive inhibitor of α-galactosidase (IC50 = 40 nM); binds to α-galactosidase in cells and chaperones unstable enzyme variants through the endoplasmic reticulum, allowing its movement into lysosomes
Catalog No. 50-296-8457 Supplier Cayman Chemical Supplier No. 1717910 MG
A competitive inhibitor of α-galactosidase (IC50 = 40 nM); binds to α-galactosidase in cells and chaperones unstable enzyme variants through the endoplasmic reticulum, allowing its movement into lysosomes