Learn More
Cayman Chemical 1-Deoxynojirimycin (hydrochloride), Purity: ≥95%, CAS: 73285-50-4, 5 mg
SDP
Catalog No. 502889213
Encompass_Preferred
An inhibitor of α-glucosidase I and II (IC50s = ~2 µM for both); inhibits virus spread in HIV-infected lymphocyte cultures at 0.4 mM
Catalog No. 50-288-9213 Supplier Cayman Chemical Supplier No. 100117185 MG
An inhibitor of α-glucosidase I and II (IC50s = ~2 µM for both); inhibits virus spread in HIV-infected lymphocyte cultures at 0.4 mM