Learn More
Cayman Chemical 1Deoxynojirimycin hydrochlor
Supplier: Cayman Chemical 1001171825 MG
An inhibitor of α-glucosidase I and II (IC50s = ~2 µM for both); inhibits virus spread in HIV-infected lymphocyte cultures at 0.4 mM