Learn More
Cayman Chemical 1-Docosahexaenoyl-2-hydroxy-sn-glycero-3-PC, Purity: ≥95% (as an 8:2 mixture of the 1-acyl and 2-acyl), CAS: 162440-05-3, 10 mg
SDP
Catalog No. 502919302
Encompass_Preferred
A lysophospholipid; has been found in shark liver extract; decreases LPS-induced increases in the levels of TNF-α and IL-6 in RAW 264.7 cells; decreases LPS-induced NO production in RAW 264.7 cells from 12-200 µM; inhibits zymosan A-induced plasma leakage into the peritoneum in a mouse model of peritonitis from 15-500 µg/kg
Catalog No. 50-291-9302 Supplier Cayman Chemical Supplier No. 3734010 MG
A lysophospholipid; has been found in shark liver extract; decreases LPS-induced increases in the levels of TNF-α and IL-6 in RAW 264.7 cells; decreases LPS-induced NO production in RAW 264.7 cells from 12-200 µM; inhibits zymosan A-induced plasma leakage into the peritoneum in a mouse model of peritonitis from 15-500 µg/kg