Learn More
Cayman Chemical 1-Docosahexaenoyl-rac-glycerol, Purity: ≥95% (mixture of isomers), CAS: 140670-40-2, 10 mg
SDP
Catalog No. 502911087
Encompass_Preferred
A monoacylglycerol; reduces ET-1-induced smooth muscle tension in isolated HPAs at 1-30 µM; reduces ET-1-induced increases in HPA cell proliferation and VEGF levels
Catalog No. 50-291-1087 Supplier Cayman Chemical Supplier No. 3587110 MG
A monoacylglycerol; reduces ET-1-induced smooth muscle tension in isolated HPAs at 1-30 µM; reduces ET-1-induced increases in HPA cell proliferation and VEGF levels