Learn More
Andwin Scientific 1DR VAIL W/TFE LINED CAP 144GR
SDP
Catalog No. NC9483432
Encompass_Preferred
The Vial Screw-Cap has a volume capacity of 4 mL. This product is constructed with borosilicate glass. It is sold in a pack of 144.
Catalog No. NC9483432 Supplier Andwin Scientific Supplier No. B69302
The Vial Screw-Cap has a volume capacity of 4 mL. This product is constructed with borosilicate glass. It is sold in a pack of 144.