Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1E,4E)-1,5-Bis[3,5-bis(methoxymethoxy)phenyl]-1,4-pentadiene-3-one 97.0+%, TCI America™
SDP

Catalog No. B4823100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4823100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4823100MG Supplier TCI America Supplier No. B4823100MG

missing translation for 'validMainframeMsg'

Specifications

CAS 917813-62-8
Melting Point 92°C
Color Yellow
Molecular Formula C25H30O9
Quantity 100 mg
Synonym GO-Y030
InChI Key WZNSZBNRYKYIQK-KQQUZDAGSA-N
SMILES COCOC1=CC(=CC(=C1)C=CC(=O)C=CC2=CC(=CC(=C2)OCOC)OCOC)OCOC
IUPAC Name (1E,4E)-1,5-bis[3,5-bis(methoxymethoxy)phenyl]penta-1,4-dien-3-one
Molecular Weight (g/mol) 474.506
PubChem CID 11641416
Formula Weight 474.51
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1E,4E)-1,5-Bis[3,5-bis(methoxymethoxy)phenyl]-1,4-pentadiene-3-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.