Learn More
Medchemexpress LLC (1E)-CFI-400437 dihydrochloride | 1247000-76-5 | 98.0% | 565.49 | 5 MG
SDP
Catalog No. 5000364248
Encompass_Preferred
(1E)-CFI-400437 dihydrochloride is a potent PLK4 inhibitor with high selectivity. It also inhibits Aurora A, Aurora B, KDR, and FLT-3, demonstrating antiproliferative activity. This compound has shown effectiveness in a mouse xenograft model of tumor growth.
  • Potent PLK4 inhibitor with IC50 of 0.6 nM
  • Selective against other PLK family members
  • Inhibits Aurora A, Aurora B, KDR, and FLT-3
  • Exhibits antiproliferative activity
  • Effective in mouse xenograft models of tumor growth
  • Plasma protein binding is 99%
Catalog No. 50-003-64248 Supplier Medchemexpress LLC Supplier No. 5MGHY132135
(1E)-CFI-400437 dihydrochloride is a potent PLK4 inhibitor with high selectivity. It also inhibits Aurora A, Aurora B, KDR, and FLT-3, demonstrating antiproliferative activity. This compound has shown effectiveness in a mouse xenograft model of tumor growth.
  • Potent PLK4 inhibitor with IC50 of 0.6 nM
  • Selective against other PLK family members
  • Inhibits Aurora A, Aurora B, KDR, and FLT-3
  • Exhibits antiproliferative activity
  • Effective in mouse xenograft models of tumor growth
  • Plasma protein binding is 99%