Learn More
AdipoGen 1E-Iodo-hex-1-ene
SDP
Catalog No. 502560643
Encompass_Preferred
Chemical. CAS 16644-98-7. Formula C6H11I. MW 210.1. Synthetic. Building block for synthesis.
Catalog No. 50-256-0643 Supplier AdipoGen Supplier No. CDXI0047G001
Chemical. CAS 16644-98-7. Formula C6H11I. MW 210.1. Synthetic. Building block for synthesis.