Learn More
Caron Products And Services 1EARTH-FRIENDLY GVAPOR
Catalog No. NC2313980
Encompass_Preferred
Earth-friendly gVapor (steam-free humidity) control from ambient to 95% RH with display/readout for Caron Model: 7401 series CO2 incubators, 708L (25 CF) & 934L (33 CF) capacities. Factory installed at the time of order.
Catalog No. NC2313980 Supplier Caron Products And Services Supplier No. HUMD307
Earth-friendly gVapor (steam-free humidity) control from ambient to 95% RH with display/readout for Caron Model: 7401 series CO2 incubators, 708L (25 CF) & 934L (33 CF) capacities. Factory installed at the time of order.