Learn More
Medchemexpress LLC HY-N2577 5mg , 1F-Fructofuranosylnystose CAS:59432-60-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25775MG
Encompass_Preferred
HY-N2577 5mg 1F-Fructofuranosylnystose CAS: 59432-60-9 1F-Fructofuranosylnystose can be used in the synthesis of Fructooligosaccharides (FOSs). Fructooligosaccharides exhibit lots of beneficial effects on our health and have been used as food ingredients. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1955