Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 1-Cyclohexenylboronic acid pinacol ester, 98%, [141091-37-4], C10330 208.105 C12H21BO2

Catalog No. 50981800
Encompass_Preferred

Compound: 1-Cyclohexenylboronic acid pinacol ester

CAS#: 141091-37-4

MDL#: MFCD05663845

Background: A cyclic vinylboronic ester widely used as a coupling partner in Suzuki-Miyaura cross-coupling reactions to introduce a cyclohexenyl group into target molecules. It is a common building block in pharmaceutical and natural product synthesis.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-981-800 Supplier Frontier Specialty Chemicals Supplier No. C103301G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1-Cyclohexenylboronic acid pinacol ester

CAS#: 141091-37-4

MDL#: MFCD05663845

Background: A cyclic vinylboronic ester widely used as a coupling partner in Suzuki-Miyaura cross-coupling reactions to introduce a cyclohexenyl group into target molecules. It is a common building block in pharmaceutical and natural product synthesis.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.