Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, (1-Methyl-1H-pyrazol-5-yl)methanol, >95%, [84547-61-5], M12944 112.13 C5H8N2O

Catalog No. 5038440
Encompass_Preferred

Compound: (1-Methyl-1H-pyrazol-5-yl)methanol

CAS#: 84547-61-5

MDL#: not listed

Background: An N-methylated pyrazole methanol used as a building block in pharmaceutical research. The alcohol group provides a reactive handle for further derivatization such as oxidation or etherification of the pyrazole heterocyclic scaffold.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-384-40 Supplier Frontier Specialty Chemicals Supplier No. M129441G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: (1-Methyl-1H-pyrazol-5-yl)methanol

CAS#: 84547-61-5

MDL#: not listed

Background: An N-methylated pyrazole methanol used as a building block in pharmaceutical research. The alcohol group provides a reactive handle for further derivatization such as oxidation or etherification of the pyrazole heterocyclic scaffold.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.