Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 1-Methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, 98%, [119083-00-0], T11436 194.111 C6H5F3N2O2

Catalog No. 5039145
Encompass_Preferred

Compound: 1-Methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

CAS#: 119083-00-0

MDL#: not listed

Background: A methylated, trifluoromethylated pyrazole carboxylic acid used as a building block in agrochemical and pharmaceutical research. The carboxylic acid group allows for amide coupling and further derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-391-45 Supplier Frontier Specialty Chemicals Supplier No. T114361G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1-Methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

CAS#: 119083-00-0

MDL#: not listed

Background: A methylated, trifluoromethylated pyrazole carboxylic acid used as a building block in agrochemical and pharmaceutical research. The carboxylic acid group allows for amide coupling and further derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.