Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 1H-Pyrazole-4-boronic acid, 98%, [763120-58-7], P7938 111.89 C3H5BN2O2

Catalog No. 50982986
Encompass_Preferred

Compound: 1H-Pyrazole-4-boronic acid

CAS#: 763120-58-7

MDL#: MFCD07368251

Background: A heterocyclic boronic acid used as a coupling partner in Suzuki-Miyaura reactions to introduce pyrazole rings into target molecules. It has been used in the synthesis of PKC-theta inhibitors and other pharmaceutically relevant scaffolds in medicinal chemistry research.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-982-986 Supplier Frontier Specialty Chemicals Supplier No. P79381G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1H-Pyrazole-4-boronic acid

CAS#: 763120-58-7

MDL#: MFCD07368251

Background: A heterocyclic boronic acid used as a coupling partner in Suzuki-Miyaura reactions to introduce pyrazole rings into target molecules. It has been used in the synthesis of PKC-theta inhibitors and other pharmaceutically relevant scaffolds in medicinal chemistry research.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.