Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2,6-Dimethoxypyridine-3-boronic acid, 98%, [221006-70-8], D1665 182.97 C7H10BNO4

Catalog No. 5036729
Encompass_Preferred

Compound: 2,6-Dimethoxypyridine-3-boronic acid

CAS#: 221006-70-8

MDL#: not listed

Background: A dimethoxylated pyridine boronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical and agrochemical research. The two methoxy groups can be later cleaved to reveal reactive pyridone functionality for additional derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-367-29 Supplier Frontier Specialty Chemicals Supplier No. D16651G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2,6-Dimethoxypyridine-3-boronic acid

CAS#: 221006-70-8

MDL#: not listed

Background: A dimethoxylated pyridine boronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical and agrochemical research. The two methoxy groups can be later cleaved to reveal reactive pyridone functionality for additional derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.