Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2-Chloropyrimidine-5-carboxaldehyde, 98%, [933702-55-7], C11270 142.543 C5H3ClN2O

Catalog No. 5036173
Encompass_Preferred

Compound: 2-Chloropyrimidine-5-carboxaldehyde

CAS#: 933702-55-7

MDL#: not listed

Background: A chlorinated pyrimidine carbaldehyde used as a building block in pharmaceutical research. The chlorine substituent supports nucleophilic aromatic substitution and cross-coupling chemistry, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-361-73 Supplier Frontier Specialty Chemicals Supplier No. C112701G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Chloropyrimidine-5-carboxaldehyde

CAS#: 933702-55-7

MDL#: not listed

Background: A chlorinated pyrimidine carbaldehyde used as a building block in pharmaceutical research. The chlorine substituent supports nucleophilic aromatic substitution and cross-coupling chemistry, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.