Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2-Chlorothiazole-4-carboxylic acid, 98%, [5198-87-8], C11739 163.582 C4H2ClNO2S

Catalog No. 5036261
Encompass_Preferred

Compound: 2-Chlorothiazole-4-carboxylic acid

CAS#: 5198-87-8

MDL#: MFCD09870033

Background: A chlorinated thiazole carboxylic acid used as a building block in pharmaceutical and agrochemical research. The chlorine substituent offers a reactive site for cross-coupling, while the carboxylic acid group allows for amide coupling.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-362-61 Supplier Frontier Specialty Chemicals Supplier No. C117391G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Chlorothiazole-4-carboxylic acid

CAS#: 5198-87-8

MDL#: MFCD09870033

Background: A chlorinated thiazole carboxylic acid used as a building block in pharmaceutical and agrochemical research. The chlorine substituent offers a reactive site for cross-coupling, while the carboxylic acid group allows for amide coupling.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.