Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2-Cyanopyridine-5-boronic acid pinacol ester, 96%, [741709-63-7], C1628 230.07 C12H15BN2O2

Catalog No. 50981855
Encompass_Preferred

Compound: 2-Cyanopyridine-5-boronic acid pinacol ester

CAS#: 741709-63-7

MDL#: MFCD06657825

Background: 2-Cyanopyridine-5-boronic acid pinacol ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals, and dyestuffs. It is used in Suzuki cross-coupling reactions to build carbon-carbon bonds in complex organic molecule synthesis.

Catalog No. 50-981-855 Supplier Frontier Specialty Chemicals Supplier No. C16281G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Cyanopyridine-5-boronic acid pinacol ester

CAS#: 741709-63-7

MDL#: MFCD06657825

Background: 2-Cyanopyridine-5-boronic acid pinacol ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals, and dyestuffs. It is used in Suzuki cross-coupling reactions to build carbon-carbon bonds in complex organic molecule synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.