Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2-(Hydroxymethyl)thiazole, 95%, [14542-12-2], H11246 115.154 C4H5NOS

Catalog No. 5037093
Encompass_Preferred

Compound: 2-(Hydroxymethyl)thiazole

CAS#: 14542-12-2

MDL#: not listed

Background: A hydroxymethyl-substituted thiazole used as a building block in pharmaceutical research. The alcohol group provides a reactive handle for further derivatization such as oxidation or etherification of the thiazole heterocyclic scaffold.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-370-93 Supplier Frontier Specialty Chemicals Supplier No. H112461G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-(Hydroxymethyl)thiazole

CAS#: 14542-12-2

MDL#: not listed

Background: A hydroxymethyl-substituted thiazole used as a building block in pharmaceutical research. The alcohol group provides a reactive handle for further derivatization such as oxidation or etherification of the thiazole heterocyclic scaffold.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.