Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2-Methoxypyridine-3-boronic acid, 98%, [163105-90-6], M10873 152.94 C6H8BNO3

Catalog No. 5038172
Encompass_Preferred

Compound: 2-Methoxypyridine-3-boronic acid

CAS#: 163105-90-6

MDL#: MFCD03411572

Background: A methoxylated pyridine boronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical and agrochemical research. The methoxy group can be later cleaved to reveal a pyridone for additional heterocyclic derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-381-72 Supplier Frontier Specialty Chemicals Supplier No. M108731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Methoxypyridine-3-boronic acid

CAS#: 163105-90-6

MDL#: MFCD03411572

Background: A methoxylated pyridine boronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical and agrochemical research. The methoxy group can be later cleaved to reveal a pyridone for additional heterocyclic derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.