Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 3,5-Dichlorophenylboronic acid, 97%, [67492-50-6], D9266 190.82 C6H5BCl2O2

Catalog No. 50982260
Encompass_Preferred

Compound: 3,5-Dichlorophenylboronic acid

CAS#: 67492-50-6

MDL#: MFCD00051935

Background: 3,5-Dichlorophenylboronic acid is used as an intermediate in pharmaceutical manufacturing and plays a role in Suzuki-Miyaura cross-coupling reactions essential for developing active pharmaceutical ingredients. It is also a reactant in trifluoromethylation, intramolecular aromatic carbenoid insertion, and cyanation reactions for synthesizing aromatic nitriles. It has additionally been used to facilitate transport of ribonucleosides across liposome membranes in biophysical research.

Catalog No. 50-982-260 Supplier Frontier Specialty Chemicals Supplier No. D92661G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3,5-Dichlorophenylboronic acid

CAS#: 67492-50-6

MDL#: MFCD00051935

Background: 3,5-Dichlorophenylboronic acid is used as an intermediate in pharmaceutical manufacturing and plays a role in Suzuki-Miyaura cross-coupling reactions essential for developing active pharmaceutical ingredients. It is also a reactant in trifluoromethylation, intramolecular aromatic carbenoid insertion, and cyanation reactions for synthesizing aromatic nitriles. It has additionally been used to facilitate transport of ribonucleosides across liposome membranes in biophysical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.