Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 3-Aminophenylboronic acid monohydrate, 98%, [206658-89-1], A6087 154.96 C6H8BNO2.H2O

Catalog No. 50981068
Encompass_Preferred

Compound: 3-Aminophenylboronic acid monohydrate

CAS#: 206658-89-1

MDL#: MFCD00149554

Background: 3-Aminophenylboronic acid monohydrate is an arylboronic acid used as a reagent in Suzuki-Miyaura cross-coupling reactions. It has been used as a boronic acid-based inhibitor of bacterial beta-lactamases, which confer resistance to penicillin and cephem antibiotics. It also serves as a building block for amphiphilic glycopolymers used in glucose-sensing nanoparticles.

Catalog No. 50-981-068 Supplier Frontier Specialty Chemicals Supplier No. A60871G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Aminophenylboronic acid monohydrate

CAS#: 206658-89-1

MDL#: MFCD00149554

Background: 3-Aminophenylboronic acid monohydrate is an arylboronic acid used as a reagent in Suzuki-Miyaura cross-coupling reactions. It has been used as a boronic acid-based inhibitor of bacterial beta-lactamases, which confer resistance to penicillin and cephem antibiotics. It also serves as a building block for amphiphilic glycopolymers used in glucose-sensing nanoparticles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.