Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 3-Bromo-4-formylpyridine, 98%, [70201-43-3], B1612 186.01 C6H4BrNO

Catalog No. 50981304
Encompass_Preferred

Compound: 3-Bromo-4-formylpyridine

CAS#: 70201-43-3

MDL#: MFCD05864506

Background: A brominated pyridine carbaldehyde used as a reagent in the synthesis of substituted oxadiazole agonists targeting the bile acid receptor TGR5, a target of interest in diabetes research. It is also used more broadly as a building block in pharmaceutical intermediate synthesis.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-981-304 Supplier Frontier Specialty Chemicals Supplier No. B16121G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Bromo-4-formylpyridine

CAS#: 70201-43-3

MDL#: MFCD05864506

Background: A brominated pyridine carbaldehyde used as a reagent in the synthesis of substituted oxadiazole agonists targeting the bile acid receptor TGR5, a target of interest in diabetes research. It is also used more broadly as a building block in pharmaceutical intermediate synthesis.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.