Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 3-Bromopyridine-5-boronic acid, 97%, [452972-09-7], B1077 201.82 C5H5BBrNO2

Catalog No. 50981174
Encompass_Preferred

Compound: 3-Bromopyridine-5-boronic acid

CAS#: 452972-09-7

MDL#: MFCD02685634

Background: 3-Bromopyridine-5-boronic acid is used in the synthesis of pharmaceuticals, including analogs of (acetylamino)(dimethylpyrazol)(pyridyl)pyrimidines studied as A2A adenosine receptor antagonists for Parkinson's disease. It serves as a versatile building block in Suzuki coupling and nickel-catalyzed cross-coupling reactions, and has been used to prepare meso-substituted porphyrins and PET imaging radioligands targeting nicotinic acetylcholine receptors.

Catalog No. 50-981-174 Supplier Frontier Specialty Chemicals Supplier No. B10771G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Bromopyridine-5-boronic acid

CAS#: 452972-09-7

MDL#: MFCD02685634

Background: 3-Bromopyridine-5-boronic acid is used in the synthesis of pharmaceuticals, including analogs of (acetylamino)(dimethylpyrazol)(pyridyl)pyrimidines studied as A2A adenosine receptor antagonists for Parkinson's disease. It serves as a versatile building block in Suzuki coupling and nickel-catalyzed cross-coupling reactions, and has been used to prepare meso-substituted porphyrins and PET imaging radioligands targeting nicotinic acetylcholine receptors.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.