Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 3-Carboxy-5-nitrophenylboronic acid, 95%, [101084-81-5], C1332 210.94 C7H6BNO6

Catalog No. 50981836
Encompass_Preferred

Compound: 3-Carboxy-5-nitrophenylboronic acid

CAS#: 101084-81-5

MDL#: MFCD00757433

Background: 3-Carboxy-5-nitrophenylboronic acid is used to prepare biaryl derivatives via Suzuki-Miyaura cross-coupling with aryl and heteroaryl halides, and to prepare 3-chloro-5-nitrobenzoic acid via copper-catalyzed chlorination. It is also used for immobilization of enzymes such as glucose oxidase and acetylcholinesterase on boronic acid-activated silica surfaces, and as a linker molecule for reversible enzyme binding in micelle design.

Catalog No. 50-981-836 Supplier Frontier Specialty Chemicals Supplier No. C13321G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Carboxy-5-nitrophenylboronic acid

CAS#: 101084-81-5

MDL#: MFCD00757433

Background: 3-Carboxy-5-nitrophenylboronic acid is used to prepare biaryl derivatives via Suzuki-Miyaura cross-coupling with aryl and heteroaryl halides, and to prepare 3-chloro-5-nitrobenzoic acid via copper-catalyzed chlorination. It is also used for immobilization of enzymes such as glucose oxidase and acetylcholinesterase on boronic acid-activated silica surfaces, and as a linker molecule for reversible enzyme binding in micelle design.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.