Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 3-Chloro-2-hydroxybenzaldehyde, >98%, [1927-94-2], C2016 156.57 C7H5ClO2

Catalog No. 50981926
Encompass_Preferred

Compound: 3-Chloro-2-hydroxybenzaldehyde

CAS#: 1927-94-2

MDL#: MFCD04973581

Background: A chlorinated salicylaldehyde used as an intermediate in the synthesis of pharmaceuticals, agrochemicals, and dyes. Derivatives of this compound have also been investigated for potential antimicrobial and antioxidant properties in research settings.

Storage: Store at room temperature, protected from light, heat, and moisture.

Catalog No. 50-981-926 Supplier Frontier Specialty Chemicals Supplier No. C20161G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Chloro-2-hydroxybenzaldehyde

CAS#: 1927-94-2

MDL#: MFCD04973581

Background: A chlorinated salicylaldehyde used as an intermediate in the synthesis of pharmaceuticals, agrochemicals, and dyes. Derivatives of this compound have also been investigated for potential antimicrobial and antioxidant properties in research settings.

Storage: Store at room temperature, protected from light, heat, and moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.