Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 3-Fluoro-2-formylpyridine, 98%, [31224-43-8], F1799 125.1 C6H4FNO

Catalog No. 50982421
Encompass_Preferred

Compound: 3-Fluoro-2-formylpyridine

CAS#: 31224-43-8

MDL#: MFCD07781234

Background: A fluorinated pyridine carbaldehyde used as a key intermediate in the synthesis of taxoid anti-tumor compounds and in the preparation of antimicrobial agents effective against drug-resistant bacteria such as MRSA. It serves as a valuable building block in pharmaceutical research.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-982-421 Supplier Frontier Specialty Chemicals Supplier No. F17991G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Fluoro-2-formylpyridine

CAS#: 31224-43-8

MDL#: MFCD07781234

Background: A fluorinated pyridine carbaldehyde used as a key intermediate in the synthesis of taxoid anti-tumor compounds and in the preparation of antimicrobial agents effective against drug-resistant bacteria such as MRSA. It serves as a valuable building block in pharmaceutical research.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.