Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 3-Iodooxetane, 95%, [26272-85-5], I12795 183.976 C3H5IO

Catalog No. 5034979
Encompass_Preferred

Compound: 3-Iodooxetane

CAS#: 26272-85-5

MDL#: not listed

Background: A small saturated oxetane bearing a reactive iodide, used as an alkylating agent in pharmaceutical research. The oxetane ring is a valuable bioisostere for gem-dimethyl or carbonyl groups, and the iodide provides a highly reactive alkylation handle.

Storage: Store at 2-8°C, protected from light.

Catalog No. 50-349-79 Supplier Frontier Specialty Chemicals Supplier No. I127951G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Iodooxetane

CAS#: 26272-85-5

MDL#: not listed

Background: A small saturated oxetane bearing a reactive iodide, used as an alkylating agent in pharmaceutical research. The oxetane ring is a valuable bioisostere for gem-dimethyl or carbonyl groups, and the iodide provides a highly reactive alkylation handle.

Storage: Store at 2-8°C, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.