Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 3-(N-Boc-aminomethyl)phenylboronic acid, 98%, [199609-62-6], B9487 251.09 C12H18BNO4

Catalog No. 50981742
Encompass_Preferred

Compound: 3-(N-Boc-aminomethyl)phenylboronic acid

CAS#: 199609-62-6

MDL#: MFCD06246052

Background: A Boc-protected aminomethyl phenylboronic acid used as a building block in Suzuki-Miyaura coupling chemistry. The protected amine allows for later deprotection and further functionalization, and this compound has been applied in the development of drug candidates for cancer and diabetes research.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-981-742 Supplier Frontier Specialty Chemicals Supplier No. B94871G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-(N-Boc-aminomethyl)phenylboronic acid

CAS#: 199609-62-6

MDL#: MFCD06246052

Background: A Boc-protected aminomethyl phenylboronic acid used as a building block in Suzuki-Miyaura coupling chemistry. The protected amine allows for later deprotection and further functionalization, and this compound has been applied in the development of drug candidates for cancer and diabetes research.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.