Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 4-Amino-3-formylpyridine, 98%, [42373-30-8], A13029 122.125 C6H6N2O

Catalog No. 5034727
Encompass_Preferred

Compound: 4-Amino-3-formylpyridine

CAS#: 42373-30-8

MDL#: not listed

Background: An amino-substituted pyridine carbaldehyde used as a building block in pharmaceutical research. The primary amine allows for amide coupling, while the aldehyde group provides a reactive handle for reductive amination.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-347-27 Supplier Frontier Specialty Chemicals Supplier No. A130291G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Amino-3-formylpyridine

CAS#: 42373-30-8

MDL#: not listed

Background: An amino-substituted pyridine carbaldehyde used as a building block in pharmaceutical research. The primary amine allows for amide coupling, while the aldehyde group provides a reactive handle for reductive amination.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.