Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 4-Bromo-2-(hydroxymethyl)thiophene, 98%, [79757-77-0], B10354 193.062 C5H5BrOS

Catalog No. 50981145
Encompass_Preferred

Compound: 4-Bromo-2-(hydroxymethyl)thiophene

CAS#: 79757-77-0

MDL#: MFCD04115392

Background: A brominated thiophene bearing a hydroxymethyl group, used as a building block in pharmaceutical and materials science research. The bromine substituent offers a reactive site for cross-coupling, while the alcohol group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-981-145 Supplier Frontier Specialty Chemicals Supplier No. B103541G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Bromo-2-(hydroxymethyl)thiophene

CAS#: 79757-77-0

MDL#: MFCD04115392

Background: A brominated thiophene bearing a hydroxymethyl group, used as a building block in pharmaceutical and materials science research. The bromine substituent offers a reactive site for cross-coupling, while the alcohol group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.