Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 4-Chloro-3-cyanophenylboronic acid, 98%, [871332-95-5], B13083 181.384 C7H5BClNO2

Catalog No. 5034786
Encompass_Preferred

Compound: 4-Chloro-3-cyanophenylboronic acid

CAS#: 871332-95-5

MDL#: not listed

Background: A chlorinated, cyano-substituted phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical research. The nitrile group provides a reactive handle for further derivatization of downstream products.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-347-86 Supplier Frontier Specialty Chemicals Supplier No. B130831G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Chloro-3-cyanophenylboronic acid

CAS#: 871332-95-5

MDL#: not listed

Background: A chlorinated, cyano-substituted phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical research. The nitrile group provides a reactive handle for further derivatization of downstream products.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.