Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 4-Fluoro-2-(trifluoromethyl)phenylboronic acid, 98%, [182344-16-7], F10834 207.92 C7H5BF4O2

Catalog No. 5036882
Encompass_Preferred

Compound: 4-Fluoro-2-(trifluoromethyl)phenylboronic acid

CAS#: 182344-16-7

MDL#: MFCD08062379

Background: A fluorinated, trifluoromethylated phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical and agrochemical research. The multiple fluorine substituents can improve metabolic stability of downstream compounds.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-368-82 Supplier Frontier Specialty Chemicals Supplier No. F108341G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Fluoro-2-(trifluoromethyl)phenylboronic acid

CAS#: 182344-16-7

MDL#: MFCD08062379

Background: A fluorinated, trifluoromethylated phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical and agrochemical research. The multiple fluorine substituents can improve metabolic stability of downstream compounds.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.