Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 4-Formylindazole, 95%, [669050-70-8], F11365 146.146 C8H6N2O

Catalog No. 5036971
Encompass_Preferred

Compound: 4-Formylindazole

CAS#: 669050-70-8

MDL#: not listed

Background: An indazole carbaldehyde used as a building block in pharmaceutical research. Indazoles are privileged nitrogen heterocyclic scaffolds in drug discovery, and the aldehyde group provides a reactive handle for reductive amination.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-369-71 Supplier Frontier Specialty Chemicals Supplier No. F113651G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Formylindazole

CAS#: 669050-70-8

MDL#: not listed

Background: An indazole carbaldehyde used as a building block in pharmaceutical research. Indazoles are privileged nitrogen heterocyclic scaffolds in drug discovery, and the aldehyde group provides a reactive handle for reductive amination.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.