Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester, 98%, [850568-72-8], B4499 304.2 C17H25BO4

Catalog No. 50981653
Encompass_Preferred

Compound: 4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

CAS#: 850568-72-8

MDL#: MFCD03453055

Background: 4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester is a product for proteomics research and a boronic ester building block commonly used in Suzuki-Miyaura cross-coupling reactions important in pharmaceutical and fine chemical synthesis. It is supplied to pharmaceutical, agricultural, and life science industries as a research chemical.

Catalog No. 50-981-653 Supplier Frontier Specialty Chemicals Supplier No. B44991G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

CAS#: 850568-72-8

MDL#: MFCD03453055

Background: 4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester is a product for proteomics research and a boronic ester building block commonly used in Suzuki-Miyaura cross-coupling reactions important in pharmaceutical and fine chemical synthesis. It is supplied to pharmaceutical, agricultural, and life science industries as a research chemical.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.