Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 5-Bromobenzo[b]thiophene, 98%, [4923-87-9], B1640 213.09 C8H5BrS

Catalog No. 50981326
Encompass_Preferred

Compound: 5-Bromobenzo[b]thiophene

CAS#: 4923-87-9

MDL#: MFCD03069318

Background: A brominated benzothiophene used as a biochemical for proteomics research and as an intermediate in organic synthesis. The bromine substituent provides a reactive handle for cross-coupling reactions to build more complex benzothiophene-containing compounds.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-981-326 Supplier Frontier Specialty Chemicals Supplier No. B16401G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromobenzo[b]thiophene

CAS#: 4923-87-9

MDL#: MFCD03069318

Background: A brominated benzothiophene used as a biochemical for proteomics research and as an intermediate in organic synthesis. The bromine substituent provides a reactive handle for cross-coupling reactions to build more complex benzothiophene-containing compounds.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.