Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 5-Bromonicotinaldehyde, 98%, [113118-81-3], B1143 186.01 C6H4BrNO

Catalog No. 50981190
Encompass_Preferred

Compound: 5-Bromonicotinaldehyde

CAS#: 113118-81-3

MDL#: MFCD03265758

Background: 5-Bromonicotinaldehyde is a key intermediate in the synthesis of pharmaceutical agents, particularly drugs targeting neurological disorders. It serves as a building block in organic synthesis for the development of heterocyclic compounds, and can be used in the synthesis of naphthyridines and diazobenzenes.

Catalog No. 50-981-190 Supplier Frontier Specialty Chemicals Supplier No. B11431G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromonicotinaldehyde

CAS#: 113118-81-3

MDL#: MFCD03265758

Background: 5-Bromonicotinaldehyde is a key intermediate in the synthesis of pharmaceutical agents, particularly drugs targeting neurological disorders. It serves as a building block in organic synthesis for the development of heterocyclic compounds, and can be used in the synthesis of naphthyridines and diazobenzenes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.