Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 5-Bromothiazole, 95%, [3034-55-7], CAPOB11216 164.024 C3H2BrNS

Catalog No. 5035500
Encompass_Preferred

Compound: 5-Bromothiazole

CAS#: 3034-55-7

MDL#: not listed

Background: A brominated thiazole used as a common building block in pharmaceutical research. Thiazole scaffolds are common in medicinal chemistry, and the bromine substituent offers a reactive site for cross-coupling.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-355-00 Supplier Frontier Specialty Chemicals Supplier No. CAPOB112161G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromothiazole

CAS#: 3034-55-7

MDL#: not listed

Background: A brominated thiazole used as a common building block in pharmaceutical research. Thiazole scaffolds are common in medicinal chemistry, and the bromine substituent offers a reactive site for cross-coupling.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.