Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole, 97%, [852227-86-2], C11378 144.602 C6H9ClN2

Catalog No. 5036203
Encompass_Preferred

Compound: 5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole

CAS#: 852227-86-2

MDL#: not listed

Background: A dimethylated chloromethylpyrazole used as a building block in pharmaceutical research. The reactive benzylic-type chloride provides a handle for alkylation reactions in pyrazole-containing scaffold synthesis.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-362-03 Supplier Frontier Specialty Chemicals Supplier No. C113781G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole

CAS#: 852227-86-2

MDL#: not listed

Background: A dimethylated chloromethylpyrazole used as a building block in pharmaceutical research. The reactive benzylic-type chloride provides a handle for alkylation reactions in pyrazole-containing scaffold synthesis.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.