Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 5-Fluoro-2-methoxynicotinaldehyde, 98%, [351410-62-3], F11569 155.126 C7H6FNO2

Catalog No. 5036981
Encompass_Preferred

Compound: 5-Fluoro-2-methoxynicotinaldehyde

CAS#: 351410-62-3

MDL#: not listed

Background: A fluorinated, methoxylated pyridine carbaldehyde used as a building block in pharmaceutical research. The fluorine substituent supports nucleophilic aromatic substitution, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-369-81 Supplier Frontier Specialty Chemicals Supplier No. F115691G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Fluoro-2-methoxynicotinaldehyde

CAS#: 351410-62-3

MDL#: not listed

Background: A fluorinated, methoxylated pyridine carbaldehyde used as a building block in pharmaceutical research. The fluorine substituent supports nucleophilic aromatic substitution, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.