Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 6-Bromo-3-formylindole, 99%, [17826-04-9], B1627 224.05 C9H6BrNO

Catalog No. 50981314
Encompass_Preferred

Compound: 6-Bromo-3-formylindole

CAS#: 17826-04-9

MDL#: MFCD00792689

Background: A halogenated indole carbaldehyde used as a key intermediate in the synthesis of pharmaceutical compounds due to its reactivity in organic reactions. It also serves as a biochemical reagent in life science research and as a building block for constructing complex organic molecules.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-981-314 Supplier Frontier Specialty Chemicals Supplier No. B16271G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 6-Bromo-3-formylindole

CAS#: 17826-04-9

MDL#: MFCD00792689

Background: A halogenated indole carbaldehyde used as a key intermediate in the synthesis of pharmaceutical compounds due to its reactivity in organic reactions. It also serves as a biochemical reagent in life science research and as a building block for constructing complex organic molecules.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.