Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 6-Bromoimidazo[1,2-a]pyrimidine, 95%, [865156-68-9], B11567 198.02 C6H4BrN3

Catalog No. 5035578
Encompass_Preferred

Compound: 6-Bromoimidazo[1,2-a]pyrimidine

CAS#: 865156-68-9

MDL#: not listed

Background: A brominated imidazopyrimidine used as a building block in pharmaceutical research. Fused imidazopyrimidine scaffolds are of interest in medicinal chemistry, and the bromine substituent provides a reactive site for cross-coupling functionalization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-355-78 Supplier Frontier Specialty Chemicals Supplier No. B115671G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 6-Bromoimidazo[1,2-a]pyrimidine

CAS#: 865156-68-9

MDL#: not listed

Background: A brominated imidazopyrimidine used as a building block in pharmaceutical research. Fused imidazopyrimidine scaffolds are of interest in medicinal chemistry, and the bromine substituent provides a reactive site for cross-coupling functionalization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.